missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Molecular Probes™ Texas Red™ Sulfonyl Chloride, mixed isomers
Texas Red™ Sulfonyl Chloride, mixed isomers
£293.00 - £359.00
Specifications
Product Type | Sulfonyl Chloride |
---|---|
Content And Storage | Store in freezer (-5 to -30°C) and protect from light. |
Chemical Reactivity | Amine |
Shipping Condition | Room Temperature |
Reactive Moiety | Sulfonyl Chloride |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10414212
|
Molecular Probes™
T1905 |
10 x 1 mg |
£359.00
10mg |
Estimated Shipment: 16-09-2024
Log in to see stock. |
|||||
11580776
|
Molecular Probes™
T353 |
10 mg |
£293.00
10mg |
Estimated Shipment: 16-09-2024
Log in to see stock. |
|||||
Description
The amine-reactive Texas Red™ sulfonyl chloride can be used to can be used to create bright red-fluorescent bioconjugates with excitation/emission maxima ∼595/615 nm.Specifications
Sulfonyl Chloride | |
Amine | |
Sulfonyl Chloride | |
Classic Dyes | |
615 |
Store in freezer (-5 to -30°C) and protect from light. | |
Room Temperature | |
Texas Red™ | |
Texas Red™ | |
595 |
For Research Use Only. Not for use in diagnostic procedures.