Learn More
Texwipe™ AlphaMop™
Product Code. p-2916359 Shop All Texwipe Products
Click to view available options
Material:
Polyester
Product Type:
Cleanroom Mop
Unit Size:
Each
Product Code. 11591214
Brand: Texwipe™ TX7108
Only null left
Add to basket
Add to basket
Flat mopping system for easy and efficient cleaning of floors, walls and ceilings
Specifically designed for the needs of ultraclean production environments. Easily changeable, laundered polyester mop covers fit over a flat, foam-padded head assembly. Lightweight, durable fiberglass handle provides excellent control over swivel head and accesses hard-to-reach areas with ease.
  • Enables rapid change of wiping surfaces for adherence to strict wiping protocol ISO Class 4 cleanroom-laundered Alpha™ polyester mop cover
  • Replaceable foam pad ensures that the polyester mop cover conforms to textured floors and other surfaces
  • Maintains ultralow generation of particles and extractables Resists corrosion, abrasion and commonly used cleaning and disinfecting chemicals such as bleaches, phenols and quaternary ammonium compound
  • AlphaMop polyester replacement mopheads and foam pads available separately (150 heads and six pads per case)
  • AlphaMop mop covers presaturated with 6% IPA Honeycomb™ 10 double-knit polyester mop covers have an abrasive texture (supplied 150 covers per case)
Specifications
Product Type Cleanroom Mop
Sterility Non-sterile
Autoclavable Autoclavable
Includes 60 in. Fiberglass Handle, 15 x 8 in. Single Head Assembly, 6 Polyester Alphawipe Mop Covers
For Use With (Application) Ideal for applications requiring low particle, extractable and ionic contamination, superior for wall, floor and ceiling cleaning, designed specifically for ultraclean manufacturing environments
Material Polyester