missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Texwipe™ AlphaMop™ Replacement Heads
AlphaMop™ 15 x 8 in. (38 x 20 cm) Thermoplastic Mop Head
Brand: Texwipe™ TX7108H
This item is not returnable.
View return policy
Description
Specifications
Replacement Head | |
Not Autoclavable | |
No |
Non-sterile | |
AlphaMop | |
Thermoplastic |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Texwipe™ AlphaMop™ Replacement Heads > Non-Autoclavable
Spot an opportunity for improvement?Share a Content Correction