missing translation for 'onlineSavingsMsg'
Learn More

Texwipe™ AlphaSorb™ 10 Wipers

Product Code. 11555405 Shop All Texwipe Products
Click to view available options
Dimensions (L x W):
9 x 9 in. (22.86 x 22.86 cm)
Unit Size:
Pack of 40
This item is not returnable. View return policy

Product Code. 11555405

Brand: Texwipe™ TX1057

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High sorbency with ultralow particles in a sealed-border wipe

Presaturated

  • Superior strength
  • Soft, nonabrasive surface
  • Presaturated with 10% isopropyl alcohol/90% deionized water

Specifications

Product Type Pre-saturated Wipe
Length (English) 9 in.
Length (Metric) 22.9 cm
Width (English) 9 in.
Width (Metric) 22.9 cm
Material Polyester
Edges Sealed
Dimensions (L x W) 9 x 9 in. (22.86 x 22.86 cm)
For Use With (Application) For aseptic cleanrooms
Saturation Solution 10% Isopropyl Alcohol, 90% Deionized Water
Sterility Sterile
Show More Show Less
compliance-icons
Product Identifier
  • X40 AlphaSat with AlphaSorb 10 9'' x 9'' Polyester , Sealed-Border Wipers Pre-Wetted With 10% IPA 23
Signal Word
  • Warning
Hazard Category
  • Flammable solid Category 2
Hazard Statement
  • H228-Flammable solid.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
Supplemental information
  • MIXTURE LIST-Contains : propan-2-ol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.