Learn More
Texwipe™ ClipperMop™ Accessories and Replacements
Texwipe ClipperMop Accessories and Replacements, Polyethylene, Nylon, Tether Clips and Nylon Screws, For Use With: TX7102 and TX7103, highest quality cleanroom wipers, For pharmaceutical, biomedical, aseptic pharmaceutical compounding and medical device industries, Recommended Applications: Safety
£259.00 - £417.79
Specifications
For Use With (Application) | For pharmaceutical, biomedical, aseptic pharmaceutical compounding and medical device industries |
---|
Product Code | Brand | Material | Product Type | For Use With (Equipment) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Material | Product Type | For Use With (Equipment) | Price | Quantity & Availability | |||
16309055
|
Texwipe™
TX7102C |
Polyethylene, Nylon | Tether Clips and Screws | TX7102 and TX7103, Highest Quality Cleanroom Wipers |
£259.00
Pack of 80 |
Estimated Shipment: 29-10-2024
Log in to see stock. |
|||
17891976
|
Texwipe™
TX7111A |
Foam | Replacement Pad | Clipper Mop Cleanroom Mop TX7102, Highest Quality Cleanroom Wipers |
£348.52
Pack of 100 |
Estimated Shipment: 16-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
17959520
|
Texwipe™
TX7111B |
Foam | Replacement Pad | Clipper Mop Cleanroom Mop TX7103, Highest Quality Cleanroom Wipers |
£417.79
Pack of 100 |
Estimated Shipment: 16-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
Description
Texwipe ClipperMop Accessories and Replacements, Polyethylene, Nylon, Tether Clips and Nylon Screws, For Use With: TX7102 and TX7103, highest quality cleanroom wipers, For pharmaceutical, biomedical, aseptic pharmaceutical compounding and medical device industries, Recommended Applications: Safety
- Clip Replacement kit
- Replacement parts for TX7102, TX7103
- There are 4 clips and 2 screws/pk.
Specifications
For pharmaceutical, biomedical, aseptic pharmaceutical compounding and medical device industries |