missing translation for 'onlineSavingsMsg'
Learn More

Texwipe™ Direct Thermal Cleanroom Labels

Designed for critical environments where direct thermal printing is used

£586.00 - £950.00

Specifications

Adhesive Type Acrylic
Format Label
Material Polypropylene
For Use With (Application) Designed for critical environments where direct thermal printing is used
Quantity 1000/Pk.
View More Specs

Products 3
Product Code Brand Size (English) Size (Metric) Price Quantity & Availability  
Product Code Brand Size (English) Size (Metric) Price Quantity & Availability  
15511255
View Documents
Texwipe™
TX542D
4 x 2 in. 10.1 x 5 cm
£894.00
4 rolls
Estimated Shipment: 04-07-2025
to see stock.
Add to basket
 
15501255
View Documents
Texwipe™
TX532D
3 x 2 in. 7.62 x 5 cm
£950.00
4 rolls
Estimated Shipment: 04-07-2025
to see stock.
Add to basket
 
15591245
View Documents
Texwipe™
TX515D
1.5 x 1.5 in. 3.8 x 3.8 cm
£586.00
4 rolls
Estimated Shipment: 04-07-2025
to see stock.
Add to basket
 
Description

Description

  • Low out-gassing polypropylene made with ultra-low leachables and low particulates
  • Quick-ship pressure sensitive
  • Available with two different adhesive levels providing permanent adhesion without leaving residue
  • Tested for surface particles, cleaned and double bagged

Specifications

Specifications

Acrylic
Polypropylene
1000/Pk.
-29 to 60°C
Label
Designed for critical environments where direct thermal printing is used
-20 to 140°F
3.3 mil
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Texwipe™ Direct Thermal Cleanroom Labels

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.