missing translation for 'onlineSavingsMsg'
Learn More

Texwipe™ Isolator Cleaning Tool Replacement Handles

£25.40 - £27.80

Specifications

Product Type Replacement Handle
Color White
Autoclavable Autoclavable
For Use With (Equipment) Isolator Cleaning Tool
Material Fiberglass
View More Specs

Products 2
Product Code Brand Length (English) Handle Price Quantity & Availability  
Product Code Brand Length (English) Handle Price Quantity & Availability  
17935241
View Documents
Texwipe™
TX7120
12 in.
£25.40
Each
Please to purchase this item. Need a web account? Register with us today!
17945241
View Documents
Texwipe™
TX7121
18 in.
£27.80
Each
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

Replacement Handle
Autoclavable
Fiberglass
White
Isolator Cleaning Tool
Videos
SDS
Documents

Documents

Product Certifications