Learn More
Texwipe™ Sterile Polystyrene Swabs
Texwipe’s Sterile Spun Polyester Swabs are constructed using 100% USP-grade polyester, securely bonded to the wooden or polystyrene handle by a water-based adhesive.
Brand:  Texwipe™ STX763
 View more versions of this product
Product Code. 17278879
  • £1135.00 / Pack of 500
Estimated Shipment: 28-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Environmental monitoring
  • Cleaning with compatible solutions and solvents
  • Picking up fine powders
  • Surface sampling
  • Collection and transport of bacteriological specimens
  • Specifications
    Specifications
    6.1 in.
    155 mm
    0.22 in.
    Polystyrene
    Polystyrene
    Foam
    Swab
    Medical-grade spun polyester
    5.8 mm
    Water-based
    Large