Learn More
Texwipe™ Sterile TechniSat™ Presaturated Wipers
For ISO Class 6 applications
Brand: Texwipe™ TX3217
Code : NEW
Additional Details : CAS Number : 67-63-0 Weight : 7.20000kg Transport : UN number : 3175 Chem class : 4.1 Pack group : II
Includes: Certificates of Irradiation, Compliance and Analysis.
Description
For ISO Class 6 applications
- Binder-free hydroentangled polyester/cellulose wiper presaturated with USP-grade 70% isopropyl alcohol 30% deionized water, filtered through 0.2μm filter
- Gamma irradiated to 10-6 Sterility Assurance Level in accordance with Association for Advancement of Medical Instrumentation
- Low levels of solvent extractables and particle generation
Order Info
Lot and expiration date clearly marked on each package. Packaging options: Case of 24 Flexpacks (50 wipers per Flexpack) or Flexpack of 20 wipers.
Compliance
Gamma-irradiated to a 10-6 SAL in accordance with the Association for the Advancement of Medical Instrumentation guidelines.
Specifications
Pre-saturated Wipers | |
28 cm | |
23 cm | |
11 x 9 in. (28 x 23 cm) | |
20 Wipers/Pk., 24 Pk./Cs., 480 Wipers Total/Cs. | |
Sterile |
11 in. | |
11 in. | |
Polyester Cellulose | |
Designed for cleaning and polishing equipment and environmental surfaces during and following the production flow | |
70% Isopropyl Alcohol, 30% Deionised Water |
Safety and Handling
- texwipes pre-wetted with 60% - 70% IPA
Signal Word
- Warning
Hazard Category
- Flammable solid Category 1
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H228-Flammable solid.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : propan-2-ol