Learn More
Texwipe™ Sterile Vertex™ Cleanroom Wipers
Ideal for delicate surfaces or applications
Brand: Texwipe™ TX3044P
393.00 GBP valid until 2024-07-31
Use promo code "21674" to get your promotional price.
Additional Details : CAS Number : 67-63-0 Weight : 4.45000kg Transport : UN number : 3175 Chem class : 4.1 Pack group : II
Description
70% ethyl alcohol wipers are less toxic and irritating to eyes than IPA.
- Confirm radiation dose met specified dose at time of receiving
- Meet Sterility Assurance Level of 10-6
- 100% polyester with a soft feel
- Processed on Texwipe Vertex manufacturing system
- Quarterly microbial testing by Agarose Overlay and Limulus Amebocyte Lysate (LAL) endotoxin testing
- Manufactured in an ISO Class 3 environment with a small physical footprint in a more sustainable process
Cleaning equipment and environmental surfaces; removal of culture residue from contact plates; wiping gloved hands; final wipedown after disinfection
Order Info
Triple-bagged
Compliance
Gamma irradiated in accordance with ANSI/AAM/ISO 11137; meets FDA Guideline for medical device bacterial endotoxin limit of not more than 20EU/device
Specifications
Vertex™ Wipes | |
30.5 cm | |
30.5 cm | |
ISO 9001:2000, 13485:2003, 14001:2004 and OHSAS 18001:1999 | |
12 x 12 in. (30.48 x 30.48 cm) | |
70% Denatured Ethanol and 30% Purified Water |
12 in. | |
12 in. | |
Polyester Knit | |
Sealed | |
Cleaning equipment and environmental surfaces | |
Sterile |