missing translation for 'onlineSavingsMsg'
Learn More

Ebro Electronic™ TFE 510 Core Thermometer

Precise and very robust thermometer with a big display

Brand:  Ebro Electronic™ 1340-5510A

Product Code. 11783499

  • £221.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Thermometer for various applications in food, laboratory, industial use. All thermometers includes a factory calibration certificate. The thermometer measures with thermocouple Type T sensor.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

EMC-Directive 2014/30/EU; EN 61326-1:2013; RoHS Directive 2011/65/EU; Electromagnetic Compatibility Regulations 2016 BS EN 61326-1:2013; The Restriction of the Use of Certain Hazardous Substances in Electrical and Electronic Equipment Regulations 2012; EN 13485
Applications: Cold store, Food products industry, Incoming goods inspection, Butchers, Food products laboratories, Catering, Food inspections
Thermometer, sensor, calibration certificate, instruction manual
Stainless Steel and Plastic
Type T Thermal Element
0.18 kg
None
300°C
-50°C
Factory Calibrated
Up to 5 years
±0.5°C/0.9°F
LCD
Single-use
52 mm
335 mm
TFE 510
0.1°C
Core Thermometer
572°F
-58°F
50°C to 300°C
AG 130, AG 140, AG 150, AG 160
Lithium 3 V/Typ CR 2477
120 mm
50 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers