missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TFE3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA554909
This item is not returnable.
View return policy
Description
Immunogen sequence: SKDLESRQRS LEQANRSLQL RIQELELQAQ IHGLPVPPTP GLLSLATTSA SDSLKPEQLD IEEEGRPGAA TFHVGGGPAQ NAPHQQPPAP PSDALLDLHF PSDHLGDLGD PFHLGLEDIL MEEEEGVVGG LSGGALSPLR AAS Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 92%.
The microphthalmia transcription factor/transcription factor E (MITF-TFE) family of basic helix-loop-helix leucine zipper (bHLH-Zip) transcription factors includes four family members: MITF, TFE3, TFEB and TFEC. The TEF3 protein encoded by this gene activates transcription through binding to the muE3 motif of the immunoglobulin heavy-chain enhancer. The TFEC protein forms heterodimers with the TEF3 protein and inhibits TFE3-dependent transcription activation. The TEF3 protein interacts with transcription regulators such as E2F3, SMAD3, and LEF-1, and is involved in TGF-beta-induced transcription, playing important roles in cell growth, proliferation, and osteoclast and macrophage differentiation. The TFE3 protein also activates hepatic IRS-2 gene, and induces hexokinase II (HK2) and insulin-induced gene 1 (INSIG1); it participates in insulin signaling and could be a therapeutic target for diabetes. This gene is also involved in chromosomal translocations, resulting in different fusion gene products in papillary renal cell carcinomas and alveolar soft part sarcomas, such as PRCC-TFE3, RCC17-TFE3, PSF-TFE3, NonO (p54nrb)-TFE3 and ASPL-TFE3.Specifications
TFE3 | |
Polyclonal | |
Unconjugated | |
TFE3 | |
TFE3; BHLHE33; Transcription factor E3; Class E basic helix-loop-helix protein 33 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
209446, 317376, 7030 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P19532, Q64092 | |
TFE3 | |
Recombit protein corresponding to Human TFE3 | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Research Use Only