missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TFE3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14480-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
The activation of gene transcription is a multistep process that is triggered by factors that recognize transcriptional enhancer sites in DNA. These factors work with co-activators to direct transcriptional initiation by the RNA polymerase II apparatus. The protein encoded by this gene is a subunit of the CRSP (cofactor required for SP1 activation) complex, which, along with TFIID, is required for efficient activation by SP1. This protein is also a component of other multisubunit complexes e.g. thyroid hormone receptor-(TR-) associated proteins which interact with TR and facilitate TR function on DNA templates in conjunction with initiation factors and cofactors. This protein contains a bipartite nuclear localization signal. This gene is known to escape chromosome X-inactivation.Specifications
TFE3 | |
Polyclonal | |
Unconjugated | |
TFE3 | |
bHLHe33, RCCP2, TFE3, TFEA, Transcription factor E3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
209446, 317376, 7030 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.25 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P19532, Q64092 | |
Tfe3 | |
TFE3 Fusion Protein Ag5863 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |