Learn More
Invitrogen™ TGF beta-1 (LAP) Human ELISA Kit
Sandwich ELISA Kit
Brand: Invitrogen™ BMS2065TEN
2046.76 GBP valid until 2024-09-28
Use promo code "21626" to get your promotional price.
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21626” to get your promotional price
Includes: Aluminum pouch(es) with a Microwell Plate coated with monoclonal antibody to human LAP
Description
The Human Transforming Growth Factor Beta 1/Latency Associated Peptide (Hu TGFβ1/LAP) ELISA quantitates Hu TGFβ1/LAP in human serum, plasma, buffered solution, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu TGFβ1/LAP. Principle of the method The Human TGFβ1/LAP solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency.Specifications
0.098ng/mL | |
Cell Culture Supernatant, Plasma (Heparin), Serum | |
ELISA | |
Latency Associated Peptide |
Biotin | |
Human | |
10 x 96 Tests | |
LAP |
For Research Use Only