Learn More
Tocris Bioscience™ TH 10785
OGG1 activator
Brand:  Tocris Bioscience™ 7778/10
Additional Details : Weight : 0.00970kg
 View more versions of this product
Product Code. 18746956
  • £188.98 / 10mg
Estimated Shipment: 05-08-2024
to see stock.
Description
Description
TH 10785 is an 8-oxo guanine DNA glycosylase 1 (OGG1) activator. It binds in the active site of OGG1 and increases its recruitment to oxidative DNA damage and its repair.
Specifications
Specifications
N-Cyclohexyl-2-cyclopropyl-4-quinazolinamine
10 mg
C17H21N3
1002801-51-5
Glycosylase Activators
0.98