Learn More
Thermo Scientific™ TH-641 Swinging Bucket Rotor Package for WX+ Ultraspeed Centrifuges
Process samples effectively with the TH-641 Swinging Bucket Rotor.
Brand:  Thermo Scientific™ 54295
19004.84 GBP valid until 2025-03-31
Use promo code "24119" to get your promotional price.
Product Code. 12161690
  • £24749.68 / Each
Explore more special offers
Description
Description
Process samples effectively with the Thermo Scientific™ TH-641 Swinging Bucket Rotor Package.
Ordering Information:
Order ultracrimp/clearcrimp sealing tool (03920), crimp gauge replacement (03919), extra plugs and caps (03045), tube removal tool (12826), tube racks (two) (03726) separately. Contact your Sales Representative for information.
Compatible with:
Thermo Scientific Sorvall WX Ultra Series Floor Model Centrifuge
Recommended for:
Rate zonal isolation of RNA and subcellular particles
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TH-641 Swinging Bucket Rotor Package
Swinging Bucket Rotor
287,660 x g
153.2 mm
114
14 mm
Thermo Scientific™ Sorvall™ WX ultraCentrifuges
211,333 x g
Titanium
6 x 13.2 mL
41,000 rpm
71.9 mm
89 mm
12.3 lb. (5.6 kg)
90°
135,005 x g
Product Suggestions
Product Suggestions
Videos
SDS
Documents Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ TH-641 Swinging Bucket Rotor Package for WX+ Ultraspeed Centrifuges > TH-641 Swinging Bucket Titanium Rotor Package