Learn More
Tocris Bioscience™ Thalidomide 4'-ether-PEG2-alkyne
Thalidomide-PEG2-alkyne
Brand: Tocris Bioscience™ 6686/25
Additional Details : Weight : 0.00970kg
Description
Thalidomide 4'-ether-PEG2-alkyne is a functionalized cereblon ligand for PROTAC® research and development; incorporates an E3 ligase ligand plus a PEG2 linker with alkyne terminal ready for conjugation to a target protein ligand. Part of a range of functionalized tool molecules for PROTACs R&D.This product has been recently renamed. The previous name for this product was Thalidomide - linker 5
Please contact us for SD files of our available Degrader Building Blocks.
PROTAC® is a registered trademark of Arvinas Operations, Inc., and is used under license.
Specifications
2-(2,6-Dioxo-3-piperidinyl)-4-[2-[2-(2-propyn-1-yloxy)oxy]ethoxy]-1H-isoindole-1,3(2H)dione | |
Thalidomide - linker 5 | |
C20H20N2O7 | |
Ligand |
2098487-52-4 | |
25 mg | |
400.38 | |
95% |