Learn More
Tocris Bioscience™ Thalidomide 4'-ether-PEG3-amine
Cereblon ligand with PEG linker and terminal amine for onward chemistry
Brand: Tocris Bioscience™ 6950/25
Additional Details : Weight : 0.00970kg
Description
Thalidomide 4'-ether-PEG3-amine is a functionalized cereblon ligand for PROTAC® research and development; incorporates an E3 ligase ligand plus a short PEG linker ready for conjugation to a target protein ligand. Part of a range of functionalized tool molecules for PROTAC R&D.This product has been recently renamed. The previous name for this product was Thalidomide - linker 11
Please contact us for SD files of our available Degrader Building Blocks.
PROTAC® is a registered trademark of Arvinas Operations, Inc., and is used under license.
Specifications
4-[2-[2-[2-(2-Aminoethoxy)ethoxy]ethoxy]ethoxy]-2-(2,6-dioxo-3-piperidinyl)-1 H-isoindole-1,3(2 H)-dione hydrochloride | |
Thalidomide - linker 11 | |
Degrader Building Blocks | |
95% |
2387510-82-7 | |
25 mg | |
C21N27N3O8.HCl |