missing translation for 'onlineSavingsMsg'
Learn More
Thallic trifluoroacetate, tech., Thermo Scientific Chemicals Product Code.: 10380334

Thallic trifluoroacetate, tech., Thermo Scientific Chemicals

Product Code. 10380334
100 g, Glass bottle
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10380334

Brand: Thermo Scientific Acros 420761000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23586-53-0
Molecular Formula C6F9O6Tl
Molecular Weight (g/mol) 543.42
MDL Number MFCD00000414
InChI Key PSHNNUKOUQCMSG-UHFFFAOYSA-K
Synonym thallium iii trifluoroacetate, acetic acid, trifluoro-, thallium 3+ salt, acetic acid,2,2,2-trifluoro-, thallium 3+ salt 3:1, thallium 3+ ; 2,2,2-trifluoroacetate, thallium tris trifluoroacetate, trifluoroacetic acid thallium iii, thallic trifluoroacetate, tech, thallium 3+ tritrifluoroacetate
PubChem CID 90200
IUPAC Name thallium(3+);2,2,2-trifluoroacetate
SMILES C(=O)(C(F)(F)F)[O-].C(=O)(C(F)(F)F)[O-].C(=O)(C(F)(F)F)[O-].[Tl+3]

Specifications

Color Beige
Physical Form Fine Powder
Quantity 100 g
Linear Formula (CF3COO)3Tl
Beilstein 02, II, 186
Fieser 03,286; 04,498; 05,658; 06,579; 07,365; 08,478; 10,397; 05,654
Formula Weight 543.42
Grade Technical
Infrared Spectrum Authentic
Packaging Glass bottle
Chemical Name or Material Thallic trifluoroacetate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

2 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0371404 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number420761000,420760250
Lot NumberA0416537 Certificate TypeCertificate of Analysis Date15/08/2022 Catalog Number420761000,420760250
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Thallic trifluoroacetate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H330-Fatal if inhaled.
  • H373-May cause damage to organs.
  • H411-Toxic to aquatic life with long lasting effects.
  • H300-Fatal if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thallic trifluoroacetate, tech., Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.