missing translation for 'onlineSavingsMsg'
Learn More
Thallium granules, 6mm (0.2in) & down, 99.99% (metals basis), packaged in water, Thermo Scientific Chemicals Product Code.: 11336498

Thallium granules, 6mm (0.2in) & down, 99.99% (metals basis), packaged in water, Thermo Scientific Chemicals

Product Code. 11336498
100 g
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy

Product Code. 11336498

Brand: Thermo Scientific Alfa Aesar 012131.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged in water
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-28-0
Molecular Formula Tl
Molecular Weight (g/mol) 204.38
MDL Number MFCD00134063
InChI Key BKVIYDNLLOSFOA-UHFFFAOYSA-N
Synonym ramor, thallium, metallic, thallium, elemental, unii-ad84r52xlf, thallium, soluble compounds, ad84r52xlf, thallium, elemental and soluble compounds, rod, 12.7mm 0.5in dia, granules, thalliumShow MoreShow Less
PubChem CID 5359464
ChEBI CHEBI:30440
IUPAC Name thallium
SMILES [Tl]

Specifications

Color Blue-White
UN Number UN1707
Quantity 100 g
Percent Purity 99.99%
Odor Odorless
Particle Size 6mm (0.2 in.) & down
Assay (metals basis)
Chemical Name or Material Thallium granules

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberX24K033 Certificate TypeCertificate of Analysis Date13/11/2024 Catalog Number012131.22,012131.14
Lot NumberP04K038 Certificate TypeCertificate of Analysis Date19/03/2024 Catalog Number012131.22,012131.14
Lot NumberX25J028 Certificate TypeCertificate of Analysis Date03/11/2023 Catalog Number012131.22,012131.14
Lot NumberM18I012 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number012131.22,012131.14
Lot NumberS29H053 Certificate TypeCertificate of Analysis Date17/11/2021 Catalog Number012131.22,012131.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thallium granules, 6mm (0.2in) & down, 99.99% (metals basis), packaged in water, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.