Learn More
Thallium(I) 2,4-pentanedionate, 97+%, Thermo Scientific Chemicals
£40.30 - £128.00
Chemical Identifiers
CAS | 14219-90-0 |
---|---|
Molecular Formula | C5H7O2Tl |
Molecular Weight (g/mol) | 303.489 |
MDL Number | MFCD00013506 |
InChI Key | QGQYKYKTENVDSC-BJILWQEISA-M |
Synonym | thallium i acetylacetonate, th acac, 2,4-pentanedione thallium i derivative, thallium 1+ 2e-4-oxopent-2-en-2-olate |
PubChem CID | 71311301 |
IUPAC Name | [(E)-4-oxopent-2-en-2-yl]oxythallium |
SMILES | CC(=CC(=O)C)O[Tl] |
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
14219-90-0 | |
303.489 | |
QGQYKYKTENVDSC-BJILWQEISA-M | |
71311301 | |
CC(=CC(=O)C)O[Tl] |
C5H7O2Tl | |
MFCD00013506 | |
thallium i acetylacetonate, th acac, 2,4-pentanedione thallium i derivative, thallium 1+ 2e-4-oxopent-2-en-2-olateShow More | |
[(E)-4-oxopent-2-en-2-yl]oxythallium |
Specifications
>134°C | |
Crystalline | |
MFCD00013506 | |
thallium i acetylacetonate, th acac, 2,4-pentanedione thallium i derivative, thallium 1+ 2e-4-oxopent-2-en-2-olate | |
CC(=CC(=O)C)O[Tl] | |
303.489 | |
303.5 | |
Thallium (I) 2,4-pentanedionate |
14219-90-0 | |
C5H7O2Tl | |
UN1707 | |
QGQYKYKTENVDSC-BJILWQEISA-M | |
[(E)-4-oxopent-2-en-2-yl]oxythallium | |
71311301 | |
≥97% |
Safety and Handling
GHS H Statement
H300-H330-H373
Fatal if swallowed.
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.
P260-P264b-P270-P271-P284-P301+P310-P304+P340-P310-P314-P330-P501c
H300+H330-H373
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: THALLIUM COMPOUNDS, N.O.S.
missing translation for 'einecsNumber' : 238-081-1
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.