Learn More
Thallium(I) acetate, 99.995% (metals basis)
Description
Thallium(I) acetate is used as a selective agent against gram-negative bacteria in selective media for the detection of mollicutes. It is also used in preparation of thallium derivatives. It is involved in the separation ore constituents by flotation method.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 131°C |
| Physical Form | Wet Crystalline Aggregates |
| Quantity | 25 g |
| UN Number | UN1707 |
| Beilstein | 3693914 |
| Merck Index | 14,9257 |
| Solubility Information | Soluble in water,alcoholSoluble in cold water,ethanol,chloroform and ether; Sparingly soluble in hot water; Insoluble in acetone. |
| Formula Weight | 263.42 |
| Percent Purity | 99.995% |
| Odor | Odorless |
| Show More |
Safety and Handling
- Thallium (I) acetate
- Danger
- Acute toxicity Category 1
- Acute toxicity Category 2
- LONG-TERM AQUATIC HAZARD Chronic 2
- Specific target organ toxicity after repeated exposure Category 2
- H300-Fatal if swallowed.
- H310-Fatal in contact with skin.
- H330-Fatal if inhaled.
- H373-May cause damage to organs.
- H411-Toxic to aquatic life with long lasting effects.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P310-Immediately call a POISON CENTER/doctor/.
- P362+P364-Take off contaminated clothing and wash it before reuse.
- P405-Store locked up.
- MIXTURE LIST-Contains : Thallium(I) acetate
RUO – Research Use Only
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.