missing translation for 'onlineSavingsMsg'
Learn More

Thallium(I) acetate, 99.995% (metals basis)

Product Code. p-7048881
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11341908

Brand: Thermo Scientific Alfa Aesar 011842.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 563-68-8 | C2H6O2Tl | 266.448 g/mol

Thallium(I) acetate is used as a selective agent against gram-negative bacteria in selective media for the detection of mollicutes. It is also used in preparation of thallium derivatives. It is involved in the separation ore constituents by flotation method.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 563-68-8
Molecular Formula C2H6O2Tl
Molecular Weight (g/mol) 266.448
MDL Number MFCD00013045
InChI Key CNWGLQAFFSLHRX-UHFFFAOYSA-N
Synonym thallium i acetate
PubChem CID 131675083
IUPAC Name acetic acid;molecular hydrogen;thallium
SMILES [HH].CC(=O)O.[Tl]

Specifications

Melting Point 131°C
Physical Form Wet Crystalline Aggregates
Quantity 25 g
UN Number UN1707
Beilstein 3693914
Merck Index 14,9257
Solubility Information Soluble in water,alcoholSoluble in cold water,ethanol,chloroform and ether; Sparingly soluble in hot water; Insoluble in acetone.
Formula Weight 263.42
Percent Purity 99.995%
Odor Odorless
Sensitivity Hygroscopic
Assay (metals basis)
Chemical Name or Material Thallium(I) acetate
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Thallium (I) acetate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H300-Fatal if swallowed.
  • H310-Fatal in contact with skin.
  • H330-Fatal if inhaled.
  • H373-May cause damage to organs.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P405-Store locked up.
Supplemental information
  • MIXTURE LIST-Contains : Thallium(I) acetate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.