missing translation for 'onlineSavingsMsg'
Learn More

Thallium(I) bromide, 99.999% (metals basis), Thermo Scientific Chemicals

£143.72 - £509.54

Chemical Identifiers

CAS 7789-40-4
MDL Number MFCD00011273
Synonym Thallous bromide
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11331908
View Documents
Thermo Scientific Alfa Aesar
011841.14
25 g
£143.72
25g
Please to purchase this item. Need a web account? Register with us today!
11321908
View Documents
Thermo Scientific Alfa Aesar
011841.22
100 g
£509.54
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Thallium(I) bromide is used as Chemical reagents and pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7789-40-4
MFCD00011273
Thallous bromide

Specifications

∼460°C
Powder
UN1707
Thallous bromide
284.28
Hygroscopic
(metals basis)
7789-40-4
MFCD00011273
14,9258
Practically insoluble in water
99.999%
7.5 g/mL
Thallium(I) bromide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H300-H330-H373
Fatal if swallowed.
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.

P260-P264b-P270-P271-P284-P301+P310-P304+P340-P310-P314-P330-P501c

H300+H330-H373

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: THALLIUM COMPOUNDS, N.O.S.

missing translation for 'einecsNumber' : 232-163-0

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only