missing translation for 'onlineSavingsMsg'
Learn More
Thallium(I) iodide, Puratronic™, 99.9995% (metals basis), Thermo Scientific Chemicals Product Code.: 11354638

Thallium(I) iodide, Puratronic™, 99.9995% (metals basis), Thermo Scientific Chemicals

Product Code. 11354638
10 g
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy

Product Code. 11354638

Brand: Thermo Scientific Alfa Aesar 043001.09

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7790-30-9
Molecular Formula ITl
Molecular Weight (g/mol) 331.28
MDL Number MFCD00011279
InChI Key CMJCEVKJYRZMIA-UHFFFAOYSA-M
Synonym thallium iii iodide, thallium iodide tli2, thallium iodide ous, thallium i iodide, anhydrous, ampuled under argon trace metals basis 10g, tliShow MoreShow Less
IUPAC Name λ¹-thallanylium iodide
SMILES [I-].[Tl+]

Specifications

Boiling Point 824°C
Melting Point 440°C
Color Yellow
Physical Form Powder
Quantity 10 g
UN Number UN1707
Merck Index 14,9264
Formula Weight 331.27
Percent Purity 99.9995%
Grade Puratronic™
Odor Odorless
Sensitivity Light sensitive
Density 7.1 g/mL
Assay (metals basis)
Chemical Name or Material Thallium (I) iodide
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberU31D032 Certificate TypeCertificate of Analysis Date15/01/2025 Catalog Number043001.18,043001.09,043001.36
Lot NumberP10J022 Certificate TypeCertificate of Analysis Date09/05/2023 Catalog Number043001.18,043001.09
Lot NumberM05C019 Certificate TypeCertificate of Analysis Date12/11/2021 Catalog Number043001.18,043001.09
Lot NumberS17C029 Certificate TypeCertificate of Analysis Date12/11/2021 Catalog Number043001.18,043001.09
Lot NumberQ22C022 Certificate TypeCertificate of Analysis Date12/11/2021 Catalog Number043001.18,043001.09

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thallium(I) iodide, Puratronic™, 99.9995% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.