missing translation for 'onlineSavingsMsg'
Learn More

Thallium(I) iodide, ultra dry, 99.999% (metals basis)

Product Code. 11348577
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
This item is not returnable. View return policy

Product Code. 11348577

Brand: Thermo Scientific Alfa Aesar 035722.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7790-30-9 | ITl | 331.28 g/mol

Thallium(I) iodide is used in mercury lamp to enhance its performance. It is mainly used for underwater lighting, since the blue green light produced has least absorption in water. Scintillator made by using iodide salt of thallium, sodium and cesium is utilized for radiation scintillation detectors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7790-30-9
Molecular Formula ITl
Molecular Weight (g/mol) 331.28
MDL Number MFCD00011279
InChI Key CMJCEVKJYRZMIA-UHFFFAOYSA-M
Synonym thallium iii iodide, thallium iodide tli2, thallium iodide ous, thallium i iodide, anhydrous, ampuled under argon trace metals basis 10g, tli
IUPAC Name λ¹-thallanylium iodide
SMILES [I-].[Tl+]

Specifications

Boiling Point 824°C
Melting Point 440°C
Color Yellow
Physical Form Beads
Quantity 50 g
Assay Percent Range 99.999% metal basis
Solubility Information Insoluble in water.
Formula Weight 331.27
Odor Odorless
Sensitivity Light sensitive
Density 7.1
Chemical Name or Material Thallium(I) iodide, ultra dry
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.