missing translation for 'onlineSavingsMsg'
Learn More

Thallium(I) neodecanoate, (Tl 52.5-55.5%), Thermo Scientific™

£55.05 - £221.00

Chemical Identifiers

MDL Number MFCD00070467
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11324577
View Documents
Thermo Scientific Alfa Aesar
039626.09
10 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11334577
View Documents
Thermo Scientific Alfa Aesar
039626.18
50 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

MFCD00070467

Specifications

Solid
UN1707
Tl 52.5 to 55.5%
MFCD00070467
375.61
Thallium(I) neodecanoate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H300-H330-H373
Fatal if swallowed.
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.

GHS P Statement
P260-P301+P310-P304+P340-P320-P330-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Specific treatment is urgent (see label).
Rinse mouth.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: THALLIUM COMPOUNDS, N.O.S.

SDS
Documents

Documents

Product Certifications