missing translation for 'onlineSavingsMsg'
Learn More

Thallium(I) nitrate, 99.5%

Product Code. p-3756719 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 10481671

Brand: Thermo Scientific Acros 265590250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10102-45-1 | NO3Tl | 266.38 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10102-45-1
Molecular Formula NO3Tl
Molecular Weight (g/mol) 266.38
MDL Number MFCD00011280
InChI Key FYWSTUCDSVYLPV-UHFFFAOYSA-N
Synonym thallium i nitrate, thallium 1+ nitrate, thallous nitrate, thallium mononitrate, thallium nitrate van, thallium nitrate, tlno3, unii-ljq38dsr12, rcra waste number u217, thallium i nitrate 1:1
PubChem CID 24937
IUPAC Name thallium(1+);nitrate
SMILES [Tl+].[O-][N+]([O-])=O

Specifications

Boiling Point 433.0°C
Melting Point 206.0°C
Color White
Physical Form Crystalline Powder
Quantity 25 g
Assay Percent Range 99.5%
Linear Formula TlNO3
Fieser 13,86
Merck Index 15, 9414
Solubility Information Solubility in water: 95g/L (20°C). Other solubilities: insoluble in alcohol
Formula Weight 266.38
Percent Purity 99.5%
Packaging Glass bottle
Chemical Name or Material Thallium(I) nitrate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.