missing translation for 'onlineSavingsMsg'
Learn More

Thallium(I) sulfate, 99.5% min (metals basis), Thermo Scientific Chemicals

Product Code. 11303918
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Thallium(I) sulfate is used as a precursor to thallium(I) sulfide, which exhibits high electrical conductivity when exposed to infrared light. It is further used in photocells. It is also mixed with selenium and arsenic to produce low melting glasses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7446-18-6
Molecular Formula O4STl2
Molecular Weight (g/mol) 504.816
MDL Number MFCD00011278
InChI Key YTQVHRVITVLIRD-UHFFFAOYSA-L
Synonym thallous sulfate, thallium sulfate, thallium i sulfate, tharattin, zelio, bonide antzix, th-universal, unii-u9f9qir12t, rcra waste number p115, sulfuric acid, dithallium 1+ salt
PubChem CID 24833
ChEBI CHEBI:81836
IUPAC Name thallium(1+);sulfate
SMILES [O-]S(=O)(=O)[O-].[Tl+].[Tl+]

Specifications

Boiling Point (decomposition)
Melting Point 632°C
Physical Form Crystalline
Quantity 50 g
Assay Percent Range 99.5% min (metals basis)
Formula Weight 504.8
Density 6.77 g/mL
Chemical Name or Material Thallium(I) sulfate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'