missing translation for 'onlineSavingsMsg'
Learn More

Thallium(I) sulfate, 99.9+%, (trace metal basis), Thermo Scientific Chemicals

Product Code. 11984241
Click to view available options
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
Plastic bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7446-18-6
Molecular Formula O4STl2
Molecular Weight (g/mol) 504.79
MDL Number MFCD00011278
InChI Key YTQVHRVITVLIRD-UHFFFAOYSA-L
Synonym thallous sulfate, thallium sulfate, thallium i sulfate, tharattin, zelio, bonide antzix, th-universal, unii-u9f9qir12t, rcra waste number p115, sulfuric acid, dithallium 1+ salt
PubChem CID 24833
ChEBI CHEBI:81836
IUPAC Name thallium(1+);sulfate
SMILES [O-]S(=O)(=O)[O-].[Tl+].[Tl+]

Specifications

Melting Point 632.0°C
Color White
Physical Form Crystals or Powder
Quantity 250 g
Assay Percent Range 80.0 to 84% (Tl)
Linear Formula TI2SO4
Merck Index 15,9418
Solubility Information Solubility in water: 4.87g/100mL (20°C)
Formula Weight 504.79
Percent Purity ≥99.9%
Grade Trace Metal Basis
Packaging Plastic bottle
Specific Gravity 6.77
Density 6.7700g/mL
Chemical Name or Material Thallium(I) sulfate, 99.9+%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Thallium(I) sulfate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H373-May cause damage to organs.
  • H411-Toxic to aquatic life with long lasting effects.
  • H300-Fatal if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.