Learn More
Thallium(III) nitrate trihydrate, 99.5% (metals basis), Thermo Scientific Chemicals
£53.06 - £893.45
Chemical Identifiers
CAS | 13453-38-8 |
---|---|
Molecular Formula | H12N3O15Tl |
Molecular Weight (g/mol) | 498.48 |
MDL Number | MFCD00149874 |
InChI Key | TWTZVACKNJEUQF-UHFFFAOYSA-N |
Synonym | nitric acid,thallium 3+ salt, trihydrate 9ci, thallium trinitrate trihydrate, thallium iii nitrate trihydrate |
PubChem CID | 11270721 |
SMILES | O.O.O.O.O.O.[Tl+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11399139
|
Thermo Scientific Alfa Aesar
087999.06 |
5 g |
£53.06
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11309149
|
Thermo Scientific Alfa Aesar
087999.14 |
25 g |
£230.15
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11389139
|
Thermo Scientific Alfa Aesar
087999.22 |
100 g |
£893.45
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Thallium(III) nitrate trihydrate is a reagent used for the rapid oxidation of styrene derivatives to carbonyl compounds, for the regioselective ring expansion of cyclic aralkyl ketones via oxidative rearrangement, for the selective hydrolysis of thioacetals to carbonyl compounds and for a general synthetic method to 19-norsteroids. Mediates ring contraction of cyclic ketones to a series of trans-fused 10-methyl-2-decalones.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
13453-38-8 | |
498.48 | |
TWTZVACKNJEUQF-UHFFFAOYSA-N | |
11270721 |
H12N3O15Tl | |
MFCD00149874 | |
nitric acid,thallium 3+ salt, trihydrate 9ci, thallium trinitrate trihydrate, thallium iii nitrate trihydrateShow More | |
O.O.O.O.O.O.[Tl+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O |
Specifications
102°C to 105°C | |
Crystalline | |
MFCD00149874 | |
nitric acid,thallium 3+ salt, trihydrate 9ci, thallium trinitrate trihydrate, thallium iii nitrate trihydrate | |
TWTZVACKNJEUQF-UHFFFAOYSA-N | |
thallium(3+);trinitrate;trihydrate | |
11270721 | |
99.5% | |
Hygroscopic | |
Thallium (III) nitrate trihydrate |
13453-38-8 | |
H12N3O15Tl | |
UN2727 | |
Soluble in water. (95g/L at 20°C). | |
O.O.O.O.O.O.[Tl+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O | |
498.48 | |
444.43 (390.38 Anhydrous) | |
Odorless | |
(metals basis) |
Safety and Handling
GHS H Statement
H300-H330-H272-H373
Fatal if swallowed.
Fatal if inhaled.
May intensify fire
oxidizer.
May cause damage to organs through prolonged or repeated exposure.
P210-P220-P221-P260-P264b-P270-P271-P280-P284-P301+P310-P304+P340-P310-P314-P330-P370+P378q-P501c
H272-H300+H330-H373
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: THALLIUM NITRATE
missing translation for 'einecsNumber' : 237-325-4
missing translation for 'rtecsNumber' : QV2000000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.