missing translation for 'onlineSavingsMsg'
Learn More

Thallium(III) oxide, 96%, Thermo Scientific Chemicals

Catalog Number 11385109
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
1314-32-5
O3Tl2
456.757
MFCD00011276
QTQRFJQXXUPYDI-UHFFFAOYSA-N
oxo oxothallanyloxy thallane, tl2o3, thallium iii oxide, oxo oxothallanyl oxy thallane, thallium oxide ic, thallium iii oxide trace metals basis
3579754
oxo(oxothallanyloxy)thallane
O=[Tl]O[Tl]=O
This item is not returnable. View return policy
1314-32-5
O3Tl2
456.757
MFCD00011276
QTQRFJQXXUPYDI-UHFFFAOYSA-N
oxo oxothallanyloxy thallane, tl2o3, thallium iii oxide, oxo oxothallanyl oxy thallane, thallium oxide ic, thallium iii oxide trace metals basis
3579754
oxo(oxothallanyloxy)thallane
O=[Tl]O[Tl]=O

Thallium(III) oxide is used in organic synthesis and as an analytical reagent

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Thallium(III) oxide is used in organic synthesis and as an analytical reagent

Solubility
Insoluble in water and alkalis. Decomposes in HCl and H2SO4

Notes
Keep container tightly closed. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY
Boiling Point 875°C
Melting Point 717°C
Quantity 10 g
UN Number UN1707
Merck Index 14,9268
Solubility Information Insoluble in water and alkalis. Decomposes in HCl and H2SO4
Formula Weight 456.74
Percent Purity 96%
Odor Odorless
Density 9.65
Chemical Name or Material Thallium(III) oxide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'