missing translation for 'onlineSavingsMsg'
Learn More

Thallium(III) oxide, 96%

Product Code. 11395109
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
This item is not returnable. View return policy

Product Code. 11395109

Brand: Thermo Scientific Alfa Aesar A17601.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1314-32-5 | O3Tl2 | 456.757 g/mol

Thallium(III) oxide is used in organic synthesis and as an analytical reagent

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Thallium(III) oxide is used in organic synthesis and as an analytical reagent

Solubility
Insoluble in water and alkalis. Decomposes in HCl and H2SO4

Notes
Keep container tightly closed. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1314-32-5
Molecular Formula O3Tl2
Molecular Weight (g/mol) 456.757
MDL Number MFCD00011276
InChI Key QTQRFJQXXUPYDI-UHFFFAOYSA-N
Synonym oxo oxothallanyloxy thallane, tl2o3, thallium iii oxide, oxo oxothallanyl oxy thallane, thallium oxide ic, thallium iii oxide trace metals basis
PubChem CID 3579754
IUPAC Name oxo(oxothallanyloxy)thallane
SMILES O=[Tl]O[Tl]=O

Specifications

Boiling Point 875°C
Melting Point 717°C
Quantity 50 g
UN Number UN1707
Merck Index 14,9268
Solubility Information Insoluble in water and alkalis. Decomposes in HCl and H2SO4
Formula Weight 456.74
Percent Purity 96%
Odor Odorless
Density 9.65
Chemical Name or Material Thallium(III) oxide
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.