missing translation for 'onlineSavingsMsg'
Learn More
Thallium, plasma standard solution, Specpure™ Tl 1000μg/mL, Thermo Scientific Chemicals Product Code.: 11341419

Thallium, plasma standard solution, Specpure™ Tl 1000μg/mL, Thermo Scientific Chemicals

Product Code. 11341419
100 mL
Click to view available options
Quantity:
50 mL
100 mL
500 mL
This item is not returnable. View return policy

Product Code. 11341419

Brand: Thermo Scientific Alfa Aesar 013851.AE

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD00134063

Specifications

Quantity 100 mL
UN Number UN3264
Physical Form Liquid
Concentration Tl in 5% HNO3
Grade Specpure™
Chemical Name or Material Thallium plasma standard solution, Tl 1000μg/mL

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number1544974 Certificate TypeCertificate of Analysis Date20/03/2025 Catalog Number013851.AD,013851.AP,013851.AE
Lot Number1526217 Certificate TypeCertificate of Analysis Date13/03/2025 Catalog Number013851.AD,013851.AP,013851.AE
Lot Number1526218 Certificate TypeCertificate of Analysis Date11/02/2025 Catalog Number013851.AD,013851.AP,013851.AE
Lot Number1521691 Certificate TypeCertificate of Analysis Date15/01/2025 Catalog Number013851.AD,013851.AP,013851.AE
Lot Number1502099 Certificate TypeCertificate of Analysis Date26/11/2024 Catalog Number013851.AD,013851.AP,013851.AE

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thallium, plasma standard solution, Specpure™ Tl 1000μg/mL, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.