missing translation for 'onlineSavingsMsg'
Learn More

Thallium Rod, 12.7mm (0.5in) dia., 99.9% (Metals basis), Thermo Scientific Chemicals

Catalog Number 11321729
Click to view available options
Quantity:
75 mm
150 mm
This item is not returnable. View return policy
7440-28-0
Tl
204.38
MFCD00134063
BKVIYDNLLOSFOA-UHFFFAOYSA-N
ramor, thallium, metallic, thallium, elemental, unii-ad84r52xlf, thallium, soluble compounds, ad84r52xlf, thallium, elemental and soluble compounds, rod, 12.7mm 0.5in dia, granules, thallium
5359464
CHEBI:30440
thallium
[Tl]
This item is not returnable. View return policy
7440-28-0
Tl
204.38
MFCD00134063
BKVIYDNLLOSFOA-UHFFFAOYSA-N
ramor, thallium, metallic, thallium, elemental, unii-ad84r52xlf, thallium, soluble compounds, ad84r52xlf, thallium, elemental and soluble compounds, rod, 12.7mm 0.5in dia, granules, thallium
5359464
CHEBI:30440
thallium
[Tl]

Form: Approx. 225g/150mm

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Appearance Rod
Assay Percent Range 99.9% (Metals basis)
Quantity 75 mm
Odor Odorless
Form Rod
Chemical Name or Material Thallium

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'