missing translation for 'onlineSavingsMsg'
Learn More
Thallium rod, 12.7mm (0.5in) dia, 99.99% (metals basis), Thermo Scientific Chemicals Product Code.: 11356677

Thallium rod, 12.7mm (0.5in) dia, 99.99% (metals basis), Thermo Scientific Chemicals

Product Code. 11356677
75 mm
Click to view available options
Quantity:
75 mm
150 mm
This item is not returnable. View return policy

Product Code. 11356677

Brand: Thermo Scientific Alfa Aesar 000888.0Y

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-28-0
Molecular Formula Tl
Molecular Weight (g/mol) 204.38
MDL Number MFCD00134063
InChI Key BKVIYDNLLOSFOA-UHFFFAOYSA-N
Synonym ramor, thallium, metallic, thallium, elemental, unii-ad84r52xlf, thallium, soluble compounds, ad84r52xlf, thallium, elemental and soluble compounds, rod, 12.7mm 0.5in dia, granules, thallium
PubChem CID 5359464
ChEBI CHEBI:30440
IUPAC Name thallium
SMILES [Tl]

Specifications

Quantity 75 mm
Percent Purity 99.99%
Odor Odorless
Weight ≈225g/150mm
Form Rod
Assay (metals basis)
Chemical Name or Material Thallium rod, 12.7mm (0.5 in.) dia.
compliance-icons compliance-icons
Product Identifier
  • Thallium rod
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Specific target organ toxicity after repeated exposure Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 4
Hazard Statement
  • H300-Fatal if swallowed.
  • H330-Fatal if inhaled.
  • H373-May cause damage to organs.
  • H413-May cause long lasting harmful effects to aquatic life.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P320-Specific treatment is urgent (see on this label).
  • P405-Store locked up.
  • P501-Dispose of contents/container to .

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thallium rod, 12.7mm (0.5in) dia, 99.99% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.