missing translation for 'onlineSavingsMsg'
Learn More

Thallium rod, 12.7mm (0.5in) dia, 99.999% (metals basis), Thermo Scientific Chemicals

£1704.00 - £2797.92

Chemical Identifiers

CAS 7440-28-0
Molecular Formula Tl
Molecular Weight (g/mol) 204.38
MDL Number MFCD00134063
InChI Key BKVIYDNLLOSFOA-UHFFFAOYSA-N
Synonym ramor, thallium, metallic, thallium, elemental, unii-ad84r52xlf, thallium, soluble compounds, ad84r52xlf, thallium, elemental and soluble compounds, rod, 12.7mm 0.5in dia, granules, thallium
PubChem CID 5359464
ChEBI CHEBI:30440
IUPAC Name thallium
SMILES [Tl]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11337517
View Documents
Thermo Scientific Alfa Aesar
012714.0Y
75 mm
£1704.00
75mm
Please to purchase this item. Need a web account? Register with us today!
11327517
View Documents
Thermo Scientific Alfa Aesar
012714.DO
150 mm
£2797.92
150mm
Please to purchase this item. Need a web account? Register with us today!
Beschreibung

Beschreibung

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Spezifikation

Chemische Identifikatoren

7440-28-0
204.38
BKVIYDNLLOSFOA-UHFFFAOYSA-N
5359464
thallium
Tl
MFCD00134063
ramor, thallium, metallic, thallium, elemental, unii-ad84r52xlf, thallium, soluble compounds, ad84r52xlf, thallium, elemental and soluble compounds, rod, 12.7mm 0.5in dia, granules, thallium
CHEBI:30440
[Tl]

Spezifikation

7440-28-0
MFCD00134063
BKVIYDNLLOSFOA-UHFFFAOYSA-N
thallium
5359464
99.999%
≈225g/150mm
(metals basis)
Tl
ramor, thallium, metallic, thallium, elemental, unii-ad84r52xlf, thallium, soluble compounds, ad84r52xlf, thallium, elemental and soluble compounds, rod, 12.7mm 0.5in dia, granules, thallium
[Tl]
204.38
CHEBI:30440
Odorless
Rod
Thallium rod, 12.7mm (0.5 in.) dia.
Videos
Sicherheit und Handhabung

Sicherheit und Handhabung

GHS H Statement
H300-H330-H373
Fatal if swallowed.
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.

P260-P264b-P270-P271-P284-P301+P310-P304+P340-P310-P314-P330-P501c

H300+H330-H373

missing translation for 'einecsNumber' : 231-138-1

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Produktzertifizierungen

RUO – Research Use Only