missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thapsigargin, 97%
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifica
| Color | Colorless |
| Assay Percent Range | 96% min. (HPLC) |
| Quantity | 5 mg |
| UN Number | 2811 |
| Merck Index | 15, 9421 |
| Synonym | thapsigargin, unii-z96bq26rzd, z96bq26rzd, chembl96926, octanoic acid 3s-3alpha, 3abeta, 4alpha, 6beta, 6abeta, 7beta, 8alpha z , 9balpha-6-acetyloxy-2,3,-3a,4,5,6,6a,7,8,9b-decahydro-3,3a-dihydroxy-3,6,9-trimethyl-8-2-methyl-1-oxo-2-butenyl oxy-2-oxo-4-1-oxobutoxy-azuleno 4,5-b furan-7-yl ester, octanoic acid, 6-acetyloxy-2,3,3a,4,5,6,6a,7,8,9b-decahydro-3,3a-dihydroxy-3,6,9-trimethyl-8-2-methyl-1-oxo-2-butenyl oxy-2-oxo-4-1-oxobutoxy azuleno 4,5-b furan-7-yl ester, 3s-3alpha,3abeta,4alpha,6beta,6abeta,7beta,8alpha z ,9balpha, tg1, --thapsigargin, octanoic acid 3s-3alpha, 3abeta, 4alpha, 6beta,6abeta, 7beta, 8alpha z , 9balpha-6-acetyloxy-2,3,-3a,4,5,6,6a,7,8,9b-decahydro-3,3a-dihydroxy-3,6,9-trimethyl-8-2-methyl-1-oxo-2-butenyl oxy-2-oxo-4-1-oxobutoxy-azuleno 4,5-b furan-7-yl ester, octanoic acid 3s-3alpha, 3abeta, 4alpha,6beta, 6abeta, 7beta, 8alpha z , 9balpha-6-acetyloxy-2,3,-3a,4,5,6,6a,7,8,9b-decahydro-3,3a-dihydroxy-3,6,9-trimethyl-8-2-methyl-1-oxo-2-butenyl oxy-2-oxo-4-1-oxobutoxy-azuleno 4,5-b furan-7-yl ester |
| Solubility Information | Solubility in water: insoluble. Other solubilities: soluble in dmso (>1 mg/mL), ethanol (5 mg/mL),, chloroform |
| Formula Weight | 650.76 |
| Percent Purity | 97% |
| Physical Form | Film |
| Vedi altri risultati |
Sicurezza e movimentazione
Identificativo prodotto
- Thapsigargin
- Danger
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Skin sensitiser Category 1
- Specific target organ toxicity Category 3
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
RUO – Research Use Only
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto