missing translation for 'onlineSavingsMsg'
Learn More

Novex™ FlexMAP™ 3D™ Calibration Kit

Product Code. 10899530
Click to view available options
Quantity:
25 reactions
Unit Size:
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10899530

Brand: Novex™ F3DCALK25

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

The FlexMAP™ 3D™ Calibration Kit

The convenient calibration kit contains xMAP™ calibrators for MagPlex and MicroPlex classification channels and also includes the calibrator for the reporter channel. Lot specific CD is included for importing target values. The kit is intended to be used with the off plate reagent area. Stripwells are included with each kit. Each kit will provide 25 calibrations.
TRUSTED_SUSTAINABILITY

Specifications

Product Line Luminex
Content And Storage Contains 28 strip wells, FM3D Calibration Kit CD, Luminex Classification Calibrator Microspheres, MagPlex Classification Calibrator Microspheres, Luminex Reporter Calibrator Microspheres. Store at 2°C to 8°C.
Quantity 25 reactions
Shipping Condition Wet Ice
For Use With (Equipment) FlexMAP 3D System
Product Type Calibration Kit

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.