Learn More
Abeba™ The Original 7091 Shoes
Animal fats, oil and fuel resistant. Sanitized™ treated.
Brand:  Abeba™ 7091/43
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 15632528
  • £44.32 / 1 pair
Estimated Shipment: 19-09-2024
to see stock.
Add to basket
Description
Description
  • CE EN ISO 20347:2012 OB FO SRC
  • Smooth leather
  • Mint green/White
  • Slip resistance outsole sole
  • Sanitized treated
  • Heel strap foldable, adjustable
  • Leather insole
Specifications
Specifications
CE EN ISO 20347:2012 OB FO SRC
Leather
Slip-resistant
Animal fats, oil and fuel
Mint Green, White
43
7091