Learn More
SymLine The Werner Purging Manifold
Install up to 4 SafetyCaps on your purging bottle with The Werner - SymLine Europe's purging manifold. No dangers, no contamination - protects people, environment and HPLC system.
Brand: SymLine 106660
204.93 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Additional Details : Weight : 0.18000kg
Description
- Safe and easy handling: capillaries are reliably fixed.
- Solvent filters have enough space due to the wide GLS80 opening.
- THE SymLine System for Solvent Safety stays permanently closed, the safety chain remains intact during the purging process.
- PE-HD, electrostatic conductive
Specifications
HPLC / UHPLC Systems | |
Black | |
HDPE | |
1 x GLS80 (f) to 4 x GL45 (m) |
Purging Manifold | |
4 | |
Purging Bottle | |
Der Werner' Collector 4-ways for HPLC rinsing bottle GLS80, PE-HD electrostatic conductive, 4 x GL45 (m) |