missing translation for 'onlineSavingsMsg'
Learn More

Theophylline, 99+%, anhydrous, Thermo Scientific Chemicals

Product Code. 10379190
Click to view available options
Quantity:
5 g
250 g
1 kg
Packaging:
Glass bottle
Plastic bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 58-55-9
Molecular Formula C7H8N4O2
Molecular Weight (g/mol) 180.17
MDL Number MFCD00079619
InChI Key ZFXYFBGIUFBOJW-UHFFFAOYSA-N
Synonym theophylline, 1,3-dimethylxanthine, elixophyllin, theophyllin, theolair, nuelin, theophylline anhydrous, respbid, theocin, theo-dur
PubChem CID 2153
ChEBI CHEBI:28177
IUPAC Name 1,3-dimethyl-7H-purine-2,6-dione
SMILES CN1C2=C(C(=O)N(C1=O)C)NC=N2

Spécification

Melting Point 270.0°C to 274.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Plastic bottle
Quantity 250 g
Merck Index 15, 9436
Solubility Information Solubility in water: 8.3g/L (20°C). Other solubilities: soluble in alkaline solutions, slightly soluble in ethanol and chloroform
Formula Weight 180.17
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material Theophylline, 99+%
Afficher plus Afficher moins
compliance-icons compliance-icons
Identifiant produit
  • Theophylline
Mention d'avertissement
  • Danger
Catégorie de danger
  • Acute toxicity Category 3
  • Reproductive toxicity Category 1B
Mention de danger
  • H301-Toxic if swallowed.
  • H360D-May damage the unborn child.
Conseils de prudence
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P405-Store locked up.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'