missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ TheraPure™ Bz dA Phosphoramidite

TheraPure™ Bz dA Phosphoramidite is a phosphoramidite containing 5' DMT and the nucleoside deoxyadenosine (dA) with Benzoyl (Bz) base protection, for use in oligonucleotide synthesis.

Brand:  Thermo Scientific™ 27-2030-75

Product Code. 15658435

  • £1750.00 / 100g

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Bz-dA CEP is a phosphoramidite containing 5′ DMT and the nucleoside deoxyadenosine (dA) with Benzoyl (Bz) base protection, for use in oligonucleotide synthesis.
  • DNA phosphoramidites are the building blocks for synthesis of DNA or DNA containing oligonucleotides.
  • Each base has unique protecting groups designed specifically to protect the DNA during the chemical synthesis process, and produce the highest amount of full length product.
  • Alternative protecting groups on dC (acetyl) and dG (DMF) allow for a faster deprotection of the oligonucleotide.
  • The TheraPure trademark denotes a highest level of phosphoramidite purity, developed with the highest stringency of of high definiteion analytics and tightly controlled impurity profiles. Ideal for any therapeutic application.
  • Features:
    • Full chemical name: 5′-O-(4,4′-Dimethoxytrityl)-N6-benzoyl-2′-deoxyadenosine-3′-O-[(2-cyanoethyl)-(N,N-diisopropyl)]-phosphoramidite
    • Molecular formula: C47H52N7O7P
    • Base: dA
    • Base Protection: Bz
    • Formula weight: 857.95 g/mol
    • CAS number: 98796-53-3
    • HPLC Purity: ≥99%; M-11 Impurity ≤0.01%; Single critical impurity ≤0.3%; Presence of dC, dG, or T impurity ≤0.03%
    • 31P NMR Purity: ≥99%; Sum of non-primary peaks at 140-152 ppm: ≤0.3%; No peak(s) above 170 ppm; HMT impurity (∼146ppm): ≤0.10%
Specifications

Specifications

5′-O-(4,4′-Dimethoxytrityl)-N6-benzoyl-2′-deoxyadenosine-3′-O-[(2-cyanoethyl)-(N,N-diisopropyl)]-phosphoramidite
857.95 g/mol
≥99%
100 g
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers