Learn More
Thermo Scientific™ TheraPure™ Modifying Enzymes
Thermo Scientific TheraPure in vitro transcription (IVT) enzymes are designed for advanced research and early development of mRNA therapeutics and vaccines.
Brand: Thermo Scientific™ EO038T1
218.04 GBP valid until 2024-11-30
Use promo code "21651" to get your promotional price.
Description
Thermo Scientific TheraPure in vitro transcription (IVT) enzymes are designed for advanced research and early development of mRNA therapeutics and vaccines. These products offer full workflow solutions from template preparation to RNA synthesis and are recommended for process research and optimization, proof of concept, and preclinical studies.
The high purity of TheraPure IVT enzymes is ensured by precise quality testing that includes confirmation that that the enzymes are nuclease- and endotoxin-free. In addition, the manufacturing process of TheraPure products follows ISO9001 and ISO13485 standards.
Features
- Convenient stand-alone enzymes for easy workflow optimization
- Animal-origin-free (AOF) raw materials used in manufacturing
- No risk of prion or animal virus contamination
- Reproducible and consistent results with validated quality testing
- Regulatory compliant—β-lactam-free manufacturing technology
- Suitable for translational research
- Facilitates transition to GMP during mRNA vaccine development process
- Scalable and available as OEM product
The exceptional quality of TheraPure products is supported by:
- Certificates of analysis
- Certificates of origin
- TSE/BSE statement
Specifications
40 U/μL | |
RNase Inhibitor | |
250 μL | |
Drug Development | |
SDS, urea, and all oxidizing reagents (p-chloromercuribenzoate, dissolved oxygen, ions) in their higher oxidation states | |
Liquid |
In vitro Transcription | |
Yes | |
RNase Inhibitor | |
TheraPure™ | |
75°C |
For Research Use Only.