missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Thermal Mixer with Blocks
Get flexibility with this vertical mixer with variable speed and temperature control for a variety of applications.
Brand: Thermo Scientific™ 13687715
742.17 GBP valid until 2024-12-20
Use promo code "21651" to get your promotional price.
This item is not returnable.
View return policy
Description
Gain variable speed and temperature control with the Thermo Scientific™ Thermal Mixer, which comes with a choice of blocks for microtubes and microplates.
- Heating and cooling capability to cover a wide range of applications
- Compact size with quiet operation
- Convenient packages that include a Thermal Mixer with a Block
- Heating and cooling capability to cover a wide range of applications
- Compact size with quiet operation
- Convenient packages that include a Thermal Mixer with a Block
Services
2 years
Specifications
20 x 5 mL (Block), 12 x 1.5 mL (Microtubes) | |
Block, 20 x 0.5 mL microtubes and 12 x 1.5 mL microtubes | |
20 x 5 mL (Block), 12 x 1.5 mL (Microtubes) |