Learn More
Thermo Scientific™ 0.2 mL Individual Tubes
0.2 mL individual tubes with attached flat or dome snap shut caps, available in a variety of colors
£47.38 - £56.34
Specifications
Certifications/Compliance | RNase, Dnase and human DNA free |
---|---|
Volume (Metric) | 0.2 mL |
Quantity | 1000 Case |
For Use With (Equipment) | 0.2mL thermal cycler blocks for PCR and sample storage applications |
Material | Polypropylene |
Product Code | Brand | Closure Type | Color | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Closure Type | Color | Price | Quantity & Availability | ||||
10592655
|
Thermo Scientific™
AB-0337/G |
Domed | Green | ||||||
10495743
|
Thermo Scientific™
AB-0337/B |
Domed | Blue | ||||||
10502855
|
Thermo Scientific™
AB-0337/R |
Domed | Red | ||||||
10502665
|
Thermo Scientific™
AB-0337/P |
Domed | Purple | ||||||
10485183
|
Thermo Scientific™
AB-0337/Y |
Domed | Yellow | ||||||
10253513
|
Thermo Scientific™
AB-0491 |
Domed Frosted | Assorted | ||||||
Description
0.2 mL Individual Tubes with integral “snap shut” caps are suitable for 0.2 mL thermal cycler blocks. They have a 0.25 mL maximum volume when closed. These individual tubes feature thin-wall design and are made from virgin polypropylene. The tubes are cleanroom produced, ensuring that they are free from contaminants that could affect experimental results. They are available in multiple colors with flat or domed caps.
The Thermo Scientific 0.2 mL Individual Tubes with integral ”snap shut“ caps are suitable for 0.2 mL thermal cycler blocks. They have a 0.25 mL maximum volume when closed. These individual tubes feature thin-wall design and are made from virgin polypropylene. The tubes are cleanroom produced, ensuring that they are free from contaminants that could affect experimental results. They are available in multiple colors with flat or domed caps.
Highlights
• Suitable for 0.2 mL thermal cycler blocks
• 0.25 mL maximum tube capacity when closed
•
Integral 'snap shut' cap
• Available with flat or domed caps
Applications
• PCR
• Sample storage
Hazardous:No
>Storage Condition:Ambient
PCR; Sample storage
Certifications
Certified free from human DNA, RNases, and DNases
Specifications
RNase, Dnase and human DNA free | |
1000 Case | |
Polypropylene | |
Thin Wall PCR Tube |
0.2 mL | |
0.2mL thermal cycler blocks for PCR and sample storage applications | |
Tube |