missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Tubes, 0.2 mL, domed cap

0.2 mL individual tubes with attached flat or dome snap shut caps, available in a variety of colors

Brand:  Thermo Scientific™ AB0337

31.39 GBP valid until 2025-04-18
Use promo code "24107" to get your promotional price.


Product Code. 10641524

  • £69.00 / Pack of 1000

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers
This item is not returnable. View return policy

Description

Description

0.2 mL Individual Tubes with integral “snap shut” caps are suitable for 0.2 mL thermal cycler blocks. They have a 0.25 mL maximum volume when closed. These individual tubes feature thin-wall design and are made from virgin polypropylene. The tubes are cleanroom produced, ensuring that they are free from contaminants that could affect experimental results. They are available in multiple colors with flat or domed caps.

The Thermo Scientific 0.2 mL Individual Tubes with integral ”snap shut“ caps are suitable for 0.2 mL thermal cycler blocks. They have a 0.25 mL maximum volume when closed. These individual tubes feature thin-wall design and are made from virgin polypropylene. The tubes are cleanroom produced, ensuring that they are free from contaminants that could affect experimental results. They are available in multiple colors with flat or domed caps.
Highlights
• Suitable for 0.2 mL thermal cycler blocks
• 0.25 mL maximum tube capacity when closed

Integral 'snap shut' cap
• Available with flat or domed caps
Applications
• PCR
• Sample storage

Hazardous:No
>Storage Condition:Ambient

PCR; Sample storage

Certifications

Certified free from human DNA, RNases, and DNases

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Certified DNA/RNase Free
0.2 mL
1000 tubes
Polypropylene
Domed Cap
Tube
Natural
0.25 mL
Thermal Cyclers
Standard
1,000 Tubes
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Tubes, 0.2 mL, domed cap > Domed Frosted Caps, Clear

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.