missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ 15% Graphite/85% Vespel Ferrules

Mechanically robust for a long lifetime and are compatible with GC/MS

Brand:  Thermo Scientific™ 290VT155

Product Code. 10688405

  • £48.05 / Pack of 10
Estimated Shipment: 08-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Form a strong grip with the column and cannot be reused as they form a permanant seal with the column
  • Temperature limit of 350°C, but must be retightened after initial temperature cycles

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Ferrules
350°C
15% Graphite, 85% Vespel
GC/MS
Thermo Scientific Trace Cold On-Column Injectors
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.