Learn More
Thermo Scientific™ Thermo Scientific™ 96-well PCR Base
Pipette with confidence using the specifically designed Thermo Scientific 96-well PCR Base in the SureTect workflow.
Brand: Thermo Scientific™ A55330
159.59 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Additional Details : Weight : 0.21000kg
Description
Easily and quickly carry out the lysis and PCR setup during the SureTect workflow using the Thermo Scientific™ 96-well PCR Base to securely hold the lysis and assay tubes. Fitting perfectly with the specifically designed Applied Biosystems™ QuantStudio™ 5 and Applied Biosystems™ SimpliAmp™ trays, the 96 well PCR Base allows confident pipetting and easy transference of tubes from rack to instrument in a single step.
Designed specifically for use with the Thermo Scientific™ SureTect™ assays, the 96-well PCR Base is one in a suite of tools enabling a simple, streamlined workflow for maximum efficiency and productivity in your laboratory.
Use the Thermo Scientific 96-well PCR Base to securely hold the lysis and assay trays and tubes when pipetting into the SureTect lysis and PCR tubes enabling a fast, easy to use workflow
Quick and Easy
Use the Thermo Scientific 96-well PCR Base to give confidence when handling and pipetting into the lysis and PCR tubes during the SureTect workflow. This tool is designed specifically for holding the SureTect tube strips during pipetting and along with the SimpliAmp and Quant Studio 5 trays enables the secure and easy handling and transferring of the SureTect lysis and assay tubes through the workflow. The rugged material and design endures constant use so can be used time and time again.Combined with our comprehensive portfolio of assays and world-class instrumentation you can perform food safety tests confidently knowing exactly what you are getting – quality combined with simplicity.
We are committed to enabling you to experience the true confidence in your results, right-first time, every time, and the Thermo Scientific™ SureTect™ Real-Time PCR System is continually evolving to support your laboratory and testing needs.
Specifications
96-well | |
96-well PCR Base | |
SureTect workflow |
Black | |
Applied Biosystems™ QuantStudio™ 5 and Applied Biosystems™ SimpliAmp™ trays | |
1 |