Learn More
Thermo Scientific™ 96-Well Separation Plate
Ready-to-use microplate format
Brand:  Thermo Scientific™ AB-0952
Product Code. 11570304
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • 0.6mL well volume
  • Contains polyethylene frits
  • Pore size 30-45μm
  • Suitable for centrifugation
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Standard
8 x 12
96